Mini bioreactor centrifuge tubes, Corning®
Lieferant: Corning
431720
734-4263EA 463.89 EUR
734-4263
Mini bioreactor centrifuge tubes, Corning®
Röhrchen Zentrifugenröhrchen
PP, translucent, with orange PP screw cap.
  • Large marking spot enables clear identification and experimental parameters
  • Easy-to-read graduations
  • Non pyrogenic
  • Free from RNase and DNase
  • Sterile
Ideal for high throughput process optimisation for suspension cell culture, the Mini Bioreactor consists of a centrifuge tube with vented cap. Tube applications include cell line developement, clone selection, media optimisation, and recombinant protein development. Screw cap features four vent holes and a 0,22 µm hydrophobic membrane to allow for gas exchange.