UV-Mikrotiterplatten, Nunc™
Lieferant: Thermo Fisher Scientific
8404
735-0253EA 788 EUR
735-0253
UV-Mikrotiterplatten, Nunc™
Mikrotiterplatten HTS-Mikrotiterplatten
Perform DNA and protein quantitation in 260 nm and 280 nm range with Thermo Scientific™ Nunc™ 96-Well UV microplates. The 96-Well format allows for a higher throughput of sample screening compared to cuvettes.
  • Non sterile
  • Flat bottom (F-well)