Multi-test verification materials, MTVM for the petroleum industry
Lieferant: STANHOPE SETA
99851-0
99853-0
99850-0
99852-0
99854-0
99908-0
751-0044EA
0
EUR
751-0044
751-0046
751-0043
751-0045
751-0047
751-0079
Multi-test verification materials, MTVM for the petroleum industry
Standards
Mineralölindustrie-Standards
Multi Test Verification Materials (MTVM) allow routine monitoring of instrument performance and are also particularly helpful in training personnel on new equipment and test methods.
- Verification to ASTM/CEN/ISO/IP and equivalent test procedures
- Cross-checks instrument performance using real product