Yttrium Standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
75592-100ML
75592-100MLEA
121
EUR
75592-100ML
Yttrium Standard for ICP-MS, TraceCERT®, Supelco®
Standards
Kationischer Standards
Yttrium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034