Auslaufbecher mit auswechselbarer Düse
Lieferant: Industrial Physics
VF2020
769-1120EA 177.29 EUR
769-1120
Auslaufbecher mit auswechselbarer Düse
Viskosimeter Auslaufbecher-Viskosimeter
Anodised aluminium cups with interchangeable stainless steel nozzles for break-thread flow time comparative measurements.
  • Engraved with a unique serial number
Typically used for the consistency of paintings and varnishes in the laboratory.
Cup weight: 226 g
Cup with orifice 4mm conforms to DIN53211
Standards: DIN 53211
Bestellinformation: Nozzles and stand must be ordered separately.