Tellurium Standard for ICP, TraceCERT®, Supelco®
Lieferant: Merck
78358-100ML
78358-100MLEA 123 EUR
78358-100ML
Tellurium Standard for ICP, TraceCERT®, Supelco®
Standards Kationischer Standards Tellur-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034