Protein Transfer buffers
Lieferant: G Bioscienes
786-019
786-019EA
222
EUR
786-019
Protein Transfer buffers
Puffer
Elektrophorese-Puffer
Transferpuffer
Efficient™ Western Transfer Buffer (20X) achieves increased protein transfer efficiency. This ready-to-use transfer buffer is prepared for optimal conductivity and efficient protein transfer, without generating excessive heat or transfer distortion.