Protein blocking solution, BLOK™ BLOTTO
Lieferant: G Bioscienes
786-192
786-192EA 299 EUR
786-192
Protein blocking solution, BLOK™ BLOTTO
Puffer Elektrophorese-Puffer Blockier-Puffer
BLOK™ BLOTTO is a 5% novel modified milk protein solution used for Westerns, ELISA and dot blot blocking.
  • Ready to use
  • Blocking in 10 to 15 mins