Protein blocking solution, BLOK™ BLOTTO
Lieferant: G Bioscienes
786-192
786-192EA
299
EUR
786-192
Protein blocking solution, BLOK™ BLOTTO
Puffer
Elektrophorese-Puffer
Blockier-Puffer
BLOK™ BLOTTO is a 5% novel modified milk protein solution used for Westerns, ELISA and dot blot blocking.
- Ready to use
- Blocking in 10 to 15 mins