Di-(N-succinimidyl)-carbonat zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate
M-Clarity™ Qualitätsstufe = MQ200
8.14971.0005
8.14971.0005EA
151
EUR
8.14971.0005
Di-(N-succinimidyl)-carbonat zur Synthese, Sigma-Aldrich®
Di(N-succinimidyl) carbonate
Formel:
C₉H₈N₂O₇ Molecular Weight: 256,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009767 CAS-Nummer: 74124-79-1 EINECS: 277-730-3 |
Spezifikation Testergebnisse
Melting Point | 188 - 191 °C |
Molar Mass | 256.17 g/mol |
Molecular weight | 256,17 g/mol |
Melting point | +188...+191 °C |
Assay (HPLC, area%) | ≥98% (a/a) |
Identity (IR) | Passes test |
Storage temperature | +15...+25 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...