n-Heptan-[D16]

Lieferant: Merck
Gefahr

Synonyme: n-Heptane-[²H₁₆]

8.15039.0005
8.15039.0005EA 0 EUR
8.15039.0005
n-Heptan-[D16]
n-Heptan-[D16]
Formel: D₃C(CD₂)₅CD₃
Molecular Weight: 116,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00062744
CAS-Nummer: 33838-52-7
UN: 1206
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR