2,7-Diaminofluoren Dihydrochlorid

Lieferant: Merck

Synonyme: 2,7-Fluorendiammoniumdichlorid

M-Clarity™ Qualitätsstufe = MQ200

8.20575.0001
8.20575.0001EA 0 EUR
8.20575.0001
2,7-Diaminofluoren Dihydrochlorid
2,7-Diaminofluoren Dihydrochlorid
Formel: C₁₃H₁₄Cl₂N₂
Molecular Weight: 269,17 g/mol
Schmelzpunkt: 260…310 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001128
CAS-Nummer: 13548-69-1
EINECS: 236-924-8
UN: 2811
ADR: 6.1,II
Merck Index: 12,04191

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Melting Point 260 - 310 °C
Molar Mass 269.17 g/mol
Synonyms 2,7-Diaminofluorene dihydrochloride
Hill Formula C13H14Cl2N2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR