Nicotinoylchlorid Hydrochlorid zur Synthese

Lieferant: Merck

8.21552.0250
8.21552.0250EA 0 EUR
8.21552.0250
Nicotinoylchlorid Hydrochlorid zur Synthese
Nicotinoylchlorid Hydrochlorid
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,02 g/mol
MDL: MFCD00012820
CAS-Nummer: 20260-53-1
EINECS: 243-648-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR