Reinraumpapier, TexWrite® 22
Lieferant: Texwipe
TX5811
818-0132EA 490 EUR
818-0132
Reinraumpapier, TexWrite® 22
Papier
TexWrite® 22, latex-saturated cellulose cleanroom documentation sheets without organic fillers.
  • Ideal for note taking, equipment manuals, and work instructions
  • Can be used with laser printers and photocopiers
Format: A4 Reinraumverbundpapier.
Verpackung: 250 Blatt pro Packung, 10 Packungen pro Karton (Bag-Within-A-Bag®).