Ethanol, BAKER, Laborreagens, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Ethyl alcohol
8229.2500PEEA
97.41
EUR
8229.2500PE
Ethanol, BAKER, Laborreagens, J.T.Baker®
Ethanol
Formel:
H₃CCH₂OH Molecular Weight: 46,07 g/mol Schmelzpunkt: –114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Spezifikation Testergebnisse
Assay (96.0-97.2% v/v)) | passes test |
Appearance | passes test |
Boiling Point | 78.0 - 79.0 °C |
Density (g/ml) at 20°C | 0.804 - 0.809 |
Iron (Fe) | max. 1 ppm |
Furfural | passes test |
Fuseloil, aldehyde | passes test |
Heavy Metals, Zinc (as Pb) | max. 2 ppm |
Identification (IR) | passes test |
Methanol (CH3OH) | max. 500 ppm |
Residue after Evaporation | max. 0.0015 % |
Substances Reducing KMnO4 | passes test |
Titrable acid | passes test |
Titrable base | passes test |
UV-absorbance (1cm vs hexane) | |
at 220 nm | max. 0.30 |
at 230 nm | max. 0.18 |
at 240 nm | max. 0.08 |
at 270 nm | max. 0.02 |
220-270 nm (no maxima) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...