Naphthol AS-MX-phosphat, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ500

855-20ML
855-20MLEA 195 EUR
855-20ML
Naphthol AS-MX-phosphat, Sigma-Aldrich®
Naphthol AS-MX-phosphat
Formel: C₁₉H₁₈NO₅P
Lagertemperatur: Gefrierschrank
MDL: MFCD00042711
CAS-Nummer: 1596-56-1
EINECS: 216-480-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR