Auramin O 85% (Farbstoffgehalt) certified by the Biological Stain Commission, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow

M-Clarity™ Qualitätsstufe = MQ200

861030-25G
861030-25GEA 52.2 EUR
861030-25G
Auramin O 85% (Farbstoffgehalt) certified by the Biological Stain Commission, Sigma-Aldrich®
Auramine O

Auramine O has been used to study the cuticle/cutin in the epidermis of the pollen presenter in Vangueria infausta. Auramine O is used for the staining of acid-fast organisms, coccidia. Auramine O along with carbol generates brilliant yellow fluorochrome of tubercle bacilli. It is effective in detecting positive cases of tuberculosis. Auramine O binds to the mycolic acid in the bacterial cell wall. Certified for use by fluorescence microscopy in Churukian′s modification of Truant′s fluorescent method for acid fast bacilli on paraffin sections.

Formel: C₁₇H₂₂ClN₃
Molecular Weight: 303,84 g/mol
Schmelzpunkt: >250 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00012484
CAS-Nummer: 2465-27-2
EINECS: 219-567-2
UN: 3077
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=964ef07339c66e4186d1e1f2a41658af6cb88440&vid=3884394008a63881a5291a170e8e4c4d27f3073f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form); Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Carbon 52.1 - 87.4 %
Nitrogen 10.7 - 18.0 %
Wavelength, c = 0.005 - 0.006 g/L; Water 429 - 435 nm
Extinction Coefficient ≥12000
Wavelength 249 - 255 nm
Solubility (Turbidity), c = 1 mg/ml; 95% Ethanol Clear to Very Hazy
Biological Stain Commission Conforms
Dye Content ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR