Xylol (Isomerengemisch) ≥98,5% (durch GC) für die Spurenanalytik von Metallen, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Dimethylbenzol

Veränderungsmanagement-Kategorie = N

8802-04EA 321.56 EUR
8802-04
Xylol (Isomerengemisch) ≥98,5% (durch GC) für die Spurenanalytik von Metallen, Macron Fine Chemicals™
Xylol (Isomerengemisch)
Formel: C₆H₄(CH₃)₂
Molecular Weight: 106,17 g/mol
Siedepunkt: 140 °C (1013 hPa)
Schmelzpunkt: –34 °C
Dichte: 0,86…0.88 g/cm³ (20 °C)
Flammpunkt: 25 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077264
CAS-Nummer: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use ≥98,5%
Assay (m,p,o-xylenes and ethylbenzene) (by GC) ≥ 98.5 %
Acidity (µeq/g) ≤ 0.3
Arsenic and Antimony (as As) ≤ 0.010 ppm
Chloride (Cl) ≤ 3 ppm
Color (APHA) ≤ 10
Heavy Metals (as Pb) ≤ 1 ppm
Identification (by IR) Passes Test
Phosphate (PO₄) ≤ 1 ppm
Residue after Evaporation ≤ 5 ppm
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤ 0.003 %
Water (H₂O) ≤ 0.02 %
Trace Impurities - Aluminum (Al) ≤ 0.100 ppm
Trace Impurities - Barium (Ba) ≤ 1.000 ppm
Trace Impurities - Boron (B) ≤ 0.100 ppm
Trace Impurities - Cadmium (Cd) ≤ 1.000 ppm
Trace Impurities - Calcium (Ca) ≤ 0.100 ppm
Trace Impurities - Chromium (Cr) ≤ 0.100 ppm
Trace Impurities - Cobalt (Co) ≤ 0.100 ppm
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
Trace Impurities - Gallium (Ga) ≤ 0.500 ppm
Trace Impurities - Germanium (Ge) ≤ 1.000 ppm
Trace Impurities - Gold (Au) ≤ 0.100 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Lead (Pb) ≤ 0.100 ppm
Trace Impurities - Lithium (Li) ≤ 1.000 ppm
Trace Impurities - Magnesium (Mg) ≤ 0.100 ppm
Trace Impurities - Manganese (Mn) ≤ 0.100 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Trace Impurities - Potassium (K) ≤ 0.100 ppm
Trace Impurities - Silicon (Si) ≤ 1.000 ppm
Trace Impurities - Silver (Ag) ≤ 0.500 ppm
Trace Impurities - Sodium (Na) ≤ 0.100 ppm
Trace Impurities - Strontium (Sr) ≤ 1.000 ppm
Trace Impurities - Tin (Sn) ≤ 0.100 ppm
Trace Impurities - Zinc (Zn) ≤ 0.100 ppm
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR