1,3-Dimethylbarbitursäure ≥98,5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
88368.180EA
123
EUR
88368.180
1,3-Dimethylbarbitursäure ≥98,5%, GPR RECTAPUR®
1,3-Dimethylbarbitursäure
Formel:
C₆H₈N₂O₃ Molecular Weight: 156,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006675 CAS-Nummer: 769-42-6 EINECS: 212-211-7 |
Spezifikation Testergebnisse
Assay (calculated on dried substance) | 98,5 - 101,5 % |
Assay (GC) (silylated) | ≥ 98,5 % |
Identification (FTIR) | Conforms |
Melting point | 120,0 - 126,0 °C |
Water | ≤ 6% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...