ortho-Phosphorsäure ≥98%, AnalaR NORMAPUR®

Lieferant: VWR Chemicals

Synonyme: Phosphorsäure, Orthophosphorsäure

89035.260EA 77 EUR
89035.260
ortho-Phosphorsäure ≥98%, AnalaR NORMAPUR®
ortho-Phosphorsäure
Formel: H₃PO₄
Molecular Weight: 98 g/mol
MDL: MFCD00011340
CAS-Nummer: 7664-38-2

Bestellen

Spezifikation Testergebnisse

Assay (GC) min. 99.9 %
Non-volatile matter max. 0.0005 %
Water (Karl Fischer) max. 0.05 %
Suitability for residue analysis max. 5 ng/l
Vapor Pressure 72.8 mmHg (20 °C)
Explosion Limit 0.16
Assay min 98.0%
Identification Passes test
Substances reducing KMnO₄ (as O) max 20 ppm
Volatile acids (as CH₃COOH) max 20 ppm
Cl (Chloride) max 1 ppm
F (Fluoride) max 0.2 ppm
NO₃ (Nitrate) max 2 ppm
PO₃ + PO₂ (as H₃PO₃) max 0.05%
SO₄ (Sulphate) max 30 ppm
As (Arsenic) max 1 ppm
Ca (Calcium) max 10 ppm
Cd (Cadmium) max 1 ppm
Co (Cobalt) max 1 ppm
Cr (Chromium) max 1 ppm
Cu (Copper) max 1 ppm
Fe (Iron) max 10 ppm
K (Potassium) max 5 ppm
Mg (Magnesium) max 1 ppm
Mn (Manganese) max 1 ppm
Na (Sodium) max 5 ppm
Ni (Nickel) max 1 ppm
Pb (Lead) max 1 ppm
Sb (Antimony) max 1 ppm
Zn (Zinc) max 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR