VWR®, Instrument Calibration Standard 2, 26 Elements in 5% HNO₃/tr. HF/tr. C₄H₆O₆
Lieferant: VWR Chemicals
91198.180EA
410
EUR
91198.180
VWR®, Instrument Calibration Standard 2, 26 Elements in 5% HNO₃/tr. HF/tr. C₄H₆O₆
Standards
Kationischer Standards
Mehrelement-Standards
26 components: 100 ug/ml each of Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn in 5% HNO₃/tr. HF/tr. C₄H₆O₆
Standards: ISO 17025/17034