Dichlormethan ≥99,5% (durch GC) inhibitor-free, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Methylenchlorid, Chlormethylen
91301-500ML
91301-500MLEA
148
EUR
91301-500ML
Dichlormethan ≥99,5% (durch GC) inhibitor-free, Selectophore™ Ionophor, Supelco®
Dichlormethan
Formel:
CH₂Cl₂ Molecular Weight: 84,93 g/mol Schmelzpunkt: −97 °C |
MDL:
MFCD00000881 CAS-Nummer: 75-09-2 UN: 1593 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...