Xylol (Isomerengemisch) ≥98,5% (durch GC) contains ethylbenzene, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Dimethylbenzol

9490-03EA 212.5 EUR
9490-03
Xylol (Isomerengemisch) ≥98,5% (durch GC) contains ethylbenzene, BAKER ANALYZED® ACS, J.T.Baker®
Xylol (Isomerengemisch)
Formel: C₆H₄(CH₃)₂
Molecular Weight: 106,17 g/mol
Siedepunkt: 140 °C (1013 hPa)
Schmelzpunkt: –34 °C
Dichte: 0,86…0.88 g/cm³ (20 °C)
Flammpunkt: 25 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077264
CAS-Nummer: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements = 98.5 %
For Laboratory,Research,or Manufacturing Use = 98.5 %
Assay (m,p,o-xylenes and ethylbenzene) (by GC) ≥ 98.5 %
Ethylbenzene (C₆H₅C₂H₅) ≤ 25 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.0020 %
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤ 0.003 %
Water (H₂O) ≤ 0.02 %
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR