D(-)-Luciferin Natriumsalz Monohydrate ≥98.5%

Lieferant: PanReac AppliChem

Synonyme: Sodium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate, D-Luciferin, sodium, D-Luciferin sodium, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt

A1006.0010
A1006.0010EA 86.7 EUR
A1006.0010
D(-)-Luciferin Natriumsalz Monohydrate ≥98.5%
D(-)-Luciferin sodium salt
Formel: C₁₁H₇N₂NaO₃S₂
Molecular Weight: 302,30 g/mol
MDL: MFCD00044938
CAS-Nummer: 103404-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR