N-Octyl-β-D-glucopyranosid

Lieferant: PanReac AppliChem
Achtung

Synonyme: Octyl-β-D-glucosid, Octyl-β-D-glucopyranosid

A1010.0025
A1010.0025EA 725 EUR
A1010.0025
N-Octyl-β-D-glucopyranosid
N-Octyl-β-D-glucopyranosid
Formel: C₁₄H₂₈O₆
Molecular Weight: 292,37 g/mol
Schmelzpunkt: 110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063288
CAS-Nummer: 29836-26-8
EINECS: 249-887-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR