L(-)-Tyrosin

Lieferant: PanReac AppliChem

Synonyme: Tyr, (S)-(-)-2-Amino-3-(4-hydroxyphenyl)propionsäure, L(-)-p-Hydroxyphenylalanin, (S)-(-)-p-Hydroxyphenylalanin, Y, H-Tyr-OH

A1677.1000
A1677.1000EA 541 EUR
A1677.1000
L(-)-Tyrosin
L(-)-Tyrosin
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Siedepunkt: 351 °C (1013 hPa)
Schmelzpunkt: 285 °C
Dichte: 1,41 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002606
CAS-Nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR