Hygromycin B
Lieferant: PanReac AppliChem
|
Gefahr
|
Synonyme:
(3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
A5347.0100
A5347.0100EA
115
EUR
A5347.0100
Hygromycin B
Hygromycin B
Formel:
C₂₀H₃₇N₃O₁₃ Molecular Weight: 527,5 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06795479 CAS-Nummer: 31282-04-9 EINECS: 250-545-5 UN: 3462 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...