Acridinorange (Chloridsalz)-Lösung 2% in Wasser, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: AO, C.I. 46005, 3,6-Bis(dimethylamino)acridine hydrochloride, Basic Orange 14, N,N,N’,N’-Tetramethyl-3,6-acridinediamine hydrochloride, 3,6-Acridinediamine, Acridine orange hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

A9231-10ML
A9231-10MLEA 145 EUR
A9231-10ML
Acridinorange (Chloridsalz)-Lösung 2% in Wasser, Sigma-Aldrich®
Acridinorange (Chloridsalz)

Acridine Orange solution.


  • hematology: suitable
  • histology: suitable


DNA intercalating dye. A grade of acridine orange of exceptionally high purity, suitable for quantitative work. Differentially stains double-stranded and single-stranded nucleic acids.

Formel: C₁₇H₁₉N₃·HCl·xH₂O
Molecular Weight: 301,81 g/mol (anhy)
Lagertemperatur: Kühlschrank
MDL: MFCD00012660
CAS-Nummer: 65-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR