iFluor® 680, Hydrazid
Lieferant: AAT BIOQUEST
Achtung
1086
AATB1086EA 307 EUR
AATB1086
iFluor® 680, Hydrazid
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d57ea64748cdc32fd8f15851d8f16f9147191a6&vid=080aab62b0531cfce479da33a2f14061ce0cc070&prid=web&clid=SAPDAM