Violet 450 acid, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1140
AATB1140EA 307 EUR
AATB1140
Violet 450 acid, mFluor™ fluoreszierende Probe
Violet 450 acid

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 542,05 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9b770381251c7297c97ed8f4f725caa00e0cba7&vid=eda9c09b3f632d4fe0dd7400633040021f8a3c34&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR