Violet 450 maleimide, mFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1600
AATB1600EA 307 EUR
AATB1600
Violet 450 maleimide, mFluor™ fluoreszierende Probe
Violet 450 maleimide

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • Large Stokes shift
  • Flow cytometer optimized
  • Stable binding

Molecular Weight: 664,63 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c200d39ada3bce0519c28aa6df7b2d776f594624&vid=f2327511f44742cdaad98ecdd8b0bf03b8e6226f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR