DAX-J2™ Orange
Lieferant: AAT BIOQUEST
|
Achtung
|
16300
AATB16300EA
230
EUR
AATB16300
DAX-J2™ Orange
DAX-J2™
DAX-J2™ Orange is a new nitric oxide (NO) sensor recently developed by AAT Bioquest.
- Does not require esterase activity for no detection
- Ph-independent fluorescence
- More photostable than daf-2 product
Bitte beachten For research use only.
Molecular Weight:
476,57 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...