ddATP [2',3'-Dideoxyadenosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Lieferant: AAT BIOQUEST
17209
AATB17209EA 292 EUR
AATB17209
ddATP [2',3'-Dideoxyadenosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Nukleinsäure-Reagenzien Reagenzien für Sequentierung
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Bitte beachten For research use only.