ReadiUse™ Mammalian Cell Lysis Buffer *5X*
Lieferant: AAT BIOQUEST
20012
AATB20012EA 152 EUR
AATB20012
ReadiUse™ Mammalian Cell Lysis Buffer *5X*
Puffer Extraktionspuffer
All our ReadiUse™ reagents require minimal hands-on time.
  • 5x concentrated
This mammalian cell lysis buffer just require a simple 5-fold dilution..