ReadiUse™ Mammalian Cell Lysis Buffer *5X*
Lieferant: AAT BIOQUEST
20012
AATB20012EA
152
EUR
AATB20012
ReadiUse™ Mammalian Cell Lysis Buffer *5X*
Puffer
Extraktionspuffer
All our ReadiUse™ reagents require minimal hands-on time.
- 5x concentrated
This mammalian cell lysis buffer just require a simple 5-fold dilution..