Dextran, M.W. 3,000 Cal-590™ labelled

Lieferant: AAT BIOQUEST

20508
AATB20508EA 460 EUR
AATB20508
Dextran, M.W. 3,000 Cal-590™ labelled
Dextran

Calcium measurement is critical for numerous biological investigations.


  • Well-retained in neurons
  • 3,000 mw
  • Useful for in vivo imaging


From Eukaryotic cells


Bitte beachten For research use only.

Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR