Dextran, M.W. 10,000 Cal-590™ labelled

Lieferant: AAT BIOQUEST

20509
AATB20509EA 460 EUR
AATB20509
Dextran, M.W. 10,000 Cal-590™ labelled
Dextran

Calcium measurement is critical for numerous biological investigations.


  • Well-retained in cells
  • 10000 mw
  • Useful for in vivo imaging


From Eukaryotic cells.


Bitte beachten For research use only.

Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR