MEQ Chlorid-Indikator

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-Methoxy-N-ethylquinolinium iodide

21250
AATB21250EA 120 EUR
AATB21250
MEQ Chlorid-Indikator
MEQ

A chloride indicator that has the fluorescence dependent of chloride concentration.


  • Chloride-dependent fluorescence
  • Well-cited

Molecular Weight: 315,15 g/mol
CAS-Nummer: 34373-76-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edeb0480e1f835bcb88331ccc5224efc08ff17de&vid=b2b5a533f89ac77d70900cd2976945ac8a0c4b27&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR