Dextran, M.W. 70,000 FITC labelled

Lieferant: AAT BIOQUEST

21702
AATB21702EA 218 EUR
AATB21702
Dextran, M.W. 70,000 FITC labelled
Dextran

FITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.


  • Well-retained in cells
  • 70,000 mw
  • Multiple applications


From Eukaryotic cells


Bitte beachten For research use only.

Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR