3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO) ≥95%

Lieferant: AAT BIOQUEST

Synonyme: DiO perchlorate, 3,3-Dioctadecyloxacarbocyanine perchlorate

22066
AATB22066EA 120 EUR
AATB22066
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO) ≥95%
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO)

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₅₃H₈₅ClN₂O₆
Molecular Weight: 881,721 g/mol
CAS-Nummer: 34215-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR