Amplite® Human Serum Albumin (HSA) Site II Binding Assay Kit
Lieferant: AAT BIOQUEST
25402
AATB25402EA
439
EUR
AATB25402
Amplite® Human Serum Albumin (HSA) Site II Binding Assay Kit
Tests
Enzymtests
Human serum albumin (HSA) is one of the most important carriers for acidic drugs in human plasma and has been shown to bind a large number of different compounds in a reversible manner.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.