iFluor® 460 styramide
Lieferant: AAT BIOQUEST
44902
AATB44902EA 429 EUR
AATB44902
iFluor® 460 styramide
iFluor® 460 styramide
iFluor® 460 Styramide is a new unique PSA reagent for multicolor application with our existing PSA and TSA reagents.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Bitte beachten For Research Use Only