iFluor® 610 styramide
Lieferant: AAT BIOQUEST
44904
AATB44904EA 429 EUR
AATB44904
iFluor® 610 styramide
iFluor® 610 styramide
iFluor® 610 Styramide is a new unique red fluorescent PSA reagent for multicolor application with our existing PSA and TSA reagents.
  • Stable for 12 months
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.