iFluor® 532 styramide
Lieferant: AAT BIOQUEST
45023
AATB45023EA 429 EUR
AATB45023
iFluor® 532 styramide
iFluor® 532 styramide
iFluor® 532 Styramide is optimized to a superior replacement for Opal Polaris 540 or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Bitte beachten For Research Use Only