iFluor® 633 styramide [Superior Replacement for Opal 650]
Lieferant: AAT BIOQUEST
45040
AATB45040EA
429
EUR
AATB45040
iFluor® 633 styramide [Superior Replacement for Opal 650]
iFluor® 633 styramide
iFluor® 633 Styramide is optimized to a superior replacement for Opal Polaris 650 or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
- Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Bitte beachten For Research Use Only