iFluor® 670 styramide [Replacement for Opal 690]
Lieferant: AAT BIOQUEST
45047
AATB45047EA
464
EUR
AATB45047
iFluor® 670 styramide [Replacement for Opal 690]
iFluor® 670 styramide
iFluor® 670 Styramide™ uses the bright red-fluorescent dye iFluor® 670 (Ex/Em = 671/682 nm) to label targets in situ.
- Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Bitte beachten For Research Use Only