SMPEG2 Proteinkonjugation
Lieferant: AAT BIOQUEST
Achtung
4510
AATB4510EA 143 EUR
AATB4510
SMPEG2 Proteinkonjugation
SMPEG2
SM(PEG)2 is an amine-to-sulfhydryl crosslinkers with 17.6 angstroms length.
  • Improved crosslinking efficiency
  • High water solubility
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=218461dad73284901a63068b7eff0cc607acb87d&vid=47c9de915217692ba26eb510a74ba4abb10df9a2&prid=web&clid=SAPDAM