SMPEG2 Proteinkonjugation
Lieferant: AAT BIOQUEST
|
Achtung
|
4510
AATB4510EA
143
EUR
AATB4510
SMPEG2 Proteinkonjugation
SMPEG2
SM(PEG)2 is an amine-to-sulfhydryl crosslinkers with 17.6 angstroms length.
- Improved crosslinking efficiency
- High water solubility
ADR:
5,II |