SMPEG4 Proteinkonjugation
Lieferant: AAT BIOQUEST
|
Achtung
|
4512
AATB4512EA
143
EUR
AATB4512
SMPEG4 Proteinkonjugation
SMPEG4
SM(PEG)4 is an amine-to-sulfhydryl crosslinkers with 24.6 angstroms length.
- Improved crosslinking efficiency
- High water solubility
ADR:
5,II |