SMPEG4 Proteinkonjugation
Lieferant: AAT BIOQUEST
Achtung
4512
AATB4512EA 143 EUR
AATB4512
SMPEG4 Proteinkonjugation
SMPEG4
SM(PEG)4 is an amine-to-sulfhydryl crosslinkers with 24.6 angstroms length.
  • Improved crosslinking efficiency
  • High water solubility
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=488df458cd0d0dc86c8783ced5baf5b539b14e6e&vid=2b7cde259f22dd91b0e6513503284ef3260842ec&prid=web&clid=SAPDAM