Polydimethylsiloxan, 100 cSt

Lieferant: ABCR

Synonyme: Silikonöl, PDMS

AB112156.0100
ABCRAB112156.0100EA 0 EUR
ABCRAB112156.0100
Polydimethylsiloxan, 100 cSt
Polydimethylsiloxane
Formel: (CH₃)₃SiO(Si(CH₃)₂O)n
Dichte: 0,965 g/cm³ (20 °C)
Flammpunkt: 321 °C (closed cup)
MDL: MFCD00084411
CAS-Nummer: 63148-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR