Polydimethylsiloxan, 10000 cSt

Lieferant: ABCR

Synonyme: Silikonöl, PDMS

AB112162.0100
ABCRAB112162.0100EA 31.9 EUR
ABCRAB112162.0100
Polydimethylsiloxan, 10000 cSt
Polydimethylsiloxane
Formel: (CH₃)₃SiO(Si(CH₃)₂O)n
MDL: MFCD00084411
CAS-Nummer: 63148-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR