Palladium(II)acetat ≥98% (99.9+%-Pd)

Lieferant: ABCR
Achtung

Synonyme: Palladiumdiacetat, Essigsäure Palladium(II)salz

AB121377.0005
ABCRAB121377.0005EA 0 EUR
ABCRAB121377.0005
Palladium(II)acetat ≥98% (99.9+%-Pd)
Palladium(II)acetat
Formel: C₄H₆O₄Pd
Molecular Weight: 224,51 g/mol
Schmelzpunkt: 205 °C
MDL: MFCD00012453
CAS-Nummer: 3375-31-3
EINECS: 222-164-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR