4-Nitrophenylchlorformiat ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Chlorameisensäure(4-nitrophenyl)ester

A18742.06 A18742.14 A18742.22 AB124931.0025
A18742.06EA 29.6 EUR
A18742.06 A18742.14 A18742.22 ABCRAB124931.0025
4-Nitrophenylchlorformiat ≥97%
4-Nitrophenylchlorformiat

Purity: 97% CAS No.: 7693-46-1 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Acrid Melting Point: 75-82° Boiling Point: 159-162°/19mm Beilstein Reference: 518127 UN No.: 3261 MDL No.: MFCD00007321

Formel: C₇H₄ClNO₄
Molecular Weight: 201,57 g/mol
Siedepunkt: 159…162 °C (19 mmHg)
Schmelzpunkt: 75…82 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00007321
CAS-Nummer: 7693-46-1
EINECS: 231-706-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR